Tisk…

1-Phenyl-2-(pyridin-4-yl)ethanone 98%

Dodavatel: Apollo Scientific

OR928199-10G OR928199-1G OR928199-5G
APOSOR928199-10GEA 24600 CZK
APOSOR928199-10G APOSOR928199-1G APOSOR928199-5G
1-Phenyl-2-(pyridin-4-yl)ethanone 98%
1-Phenyl-2-(pyridin-4-yl)ethanone
Vzorec: C₁₃H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00234969
CAS číslo: 1620-55-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR