Tisk…

9,9-Bis(methoxymethyl)fluorene 97%

Dodavatel: Apollo Scientific

OR928153-1G OR928153-250MG OR928153-5G
APOSOR928153-1GEA 3110 CZK
APOSOR928153-1G APOSOR928153-250MG APOSOR928153-5G
9,9-Bis(methoxymethyl)fluorene 97%
9,9-Bis(methoxymethyl)fluorene
Vzorec: C₁₇H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD19441380
CAS číslo: 182121-12-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR