Tisk…

2-Methoxy-4-nitrobenzyl alcohol 95%

Dodavatel: Apollo Scientific

OR928139-1G OR928139-250MG
APOSOR928139-1GEA 1910 CZK
APOSOR928139-1G APOSOR928139-250MG
2-Methoxy-4-nitrobenzyl alcohol 95%
2-Methoxy-4-nitrobenzyl alcohol
Vzorec: C₈H₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD08277262
CAS číslo: 136507-14-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR