Tisk…

Ethyl 6-formylnicotinate 95%

Dodavatel: Apollo Scientific

OR928135-1G OR928135-250MG
APOSOR928135-1GEA 23970 CZK
APOSOR928135-1G APOSOR928135-250MG
Ethyl 6-formylnicotinate 95%
Ethyl 6-formylnicotinate
Vzorec: C₉H₉NO₃
MW: 179.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19690524
CAS číslo: 20857-31-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR