2-Pyridin-4-yl-phenylamine 95%
Dodavatel: Apollo Scientific
OR928031-1G
OR928031-250MG
APOSOR928031-1GEA
15310
CZK
APOSOR928031-1G
APOSOR928031-250MG
2-Pyridin-4-yl-phenylamine 95%
2-Pyridin-4-yl-phenylamine
Vzorec:
C₁₁H₁₀N₂ MW: 170,211 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06738704 CAS číslo: 106047-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...