quinoline-4-carbothioamide 95%
Dodavatel: Apollo Scientific
OR928020-1G
OR928020-250MG
APOSOR928020-1GEA
21490
CZK
APOSOR928020-1G
APOSOR928020-250MG
quinoline-4-carbothioamide 95%
quinoline-4-carbothioamide
Vzorec:
C₁₀H₈N₂S MW: 188,249 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02179220 CAS číslo: 74585-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...