6-Chloroindole-3-acetonitrile 95%
Dodavatel: Apollo Scientific
OR927971-1G
OR927971-5G
APOSOR927971-1GEA
4570
CZK
APOSOR927971-1G
APOSOR927971-5G
6-Chloroindole-3-acetonitrile 95%
6-Chloroindole-3-acetonitrile
Vzorec:
C₁₀H₇ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD03428598 CAS číslo: 61220-58-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...