Tisk…

5-Bromo-2-methyl-2H-pyrazolo[3,4-b]pyridine 95%

Dodavatel: Apollo Scientific

OR927931-1G OR927931-250MG OR927931-5G
APOSOR927931-1GEA 2200 CZK
APOSOR927931-1G APOSOR927931-250MG APOSOR927931-5G
5-Bromo-2-methyl-2H-pyrazolo[3,4-b]pyridine 95%
5-Bromo-2-methyl-2H-pyrazolo[3,4-b]pyridine
Vzorec: C₇H₆BrN₃
MW: 212,047 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19443914
CAS číslo: 1316852-65-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR