Tisk…

Methyl-(3-phenoxy-phenyl)-amine 95%

Dodavatel: Apollo Scientific

Synonyms: N-Methyl-3-phenoxyaniline

OR927848-1G OR927848-250MG
APOSOR927848-1GEA 16340 CZK
APOSOR927848-1G APOSOR927848-250MG
Methyl-(3-phenoxy-phenyl)-amine 95%
Methyl-(3-phenoxy-phenyl)-amine
Vzorec: C₁₃H₁₃NO
MW: 199,248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738789
CAS číslo: 13024-17-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR