2-(4-chlorophenyl)acetaldehyde 95%
Dodavatel: Apollo Scientific
Synonyms:
(4-Chlorophenyl)acetaldehyde
OR927834-1G
OR927834-250MG
APOSOR927834-1GEA
6390
CZK
APOSOR927834-1G
APOSOR927834-250MG
2-(4-chlorophenyl)acetaldehyde 95%
2-(4-chlorophenyl)acetaldehyde
Vzorec:
C₈H₇ClO Storage Temperature: Freezer |
MDL Number:
MFCD02261732 CAS číslo: 4251-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...