Tisk…

Chloromethyl ethyl carbonate 97%

Dodavatel: Apollo Scientific

OR927715-1G OR927715-5G
APOSOR927715-1GEA 2200 CZK
APOSOR927715-1G APOSOR927715-5G
Chloromethyl ethyl carbonate 97%
Chloromethyl ethyl carbonate
Vzorec: C₄H₇ClO₃
MW: 138,55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00137995
CAS číslo: 35179-98-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR