Tisk…

4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 97%

Dodavatel: Apollo Scientific

OR927654-10G OR927654-1G OR927654-5G
APOSOR927654-10GEA 14680 CZK
APOSOR927654-10G APOSOR927654-1G APOSOR927654-5G
4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 97%
4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
Vzorec: C₈H₅ClN₂O
MW: 180,591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09880129
CAS číslo: 918515-16-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR