Tisk…

5-tert-Butyl-2-hydroxybenzoic acid 96%

Dodavatel: Apollo Scientific

Synonyms: 5-tert-Butyl-2-hydroxybenzoic Acid

OR927645-1G OR927645-250MG OR927645-5G
APOSOR927645-1GEA 7250 CZK
APOSOR927645-1G APOSOR927645-250MG APOSOR927645-5G
5-tert-Butyl-2-hydroxybenzoic acid 96%
5-tert-Butyl-2-hydroxybenzoic acid
Vzorec: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00156971
CAS číslo: 16094-31-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR