Tisk…

2-(1-Piperidinyl)benzoic acid 97%

Dodavatel: Apollo Scientific

OR927641-1G OR927641-5G
APOSOR927641-1GEA 7250 CZK
APOSOR927641-1G APOSOR927641-5G
2-(1-Piperidinyl)benzoic acid 97%
2-(1-Piperidinyl)benzoic acid
Vzorec: C₁₂H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00275762
CAS číslo: 42093-97-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR