Tisk…

5-Methylbenzo[d]isoxazol-3-amine 98%

Dodavatel: Apollo Scientific

OR927597-1G OR927597-250MG OR927597-5G
APOSOR927597-1GEA 8500 CZK
APOSOR927597-1G APOSOR927597-250MG APOSOR927597-5G
5-Methylbenzo[d]isoxazol-3-amine 98%
5-Methylbenzo[d]isoxazol-3-amine
Vzorec: C₈H₈N₂O
MW: 148,162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907911
CAS číslo: 89976-56-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR