Tisk…

7-Methoxyquinazoline-2,4-diol 95%

Dodavatel: Apollo Scientific

OR927579-1G OR927579-5G
APOSOR927579-1GEA 1540 CZK
APOSOR927579-1G APOSOR927579-5G
7-Methoxyquinazoline-2,4-diol 95%
7-Methoxyquinazoline-2,4-diol
Vzorec: C₉H₈N₂O₃
MW: 192,171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18427204
CAS číslo: 62484-12-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR