(4-Phenylmorpholin-2-yl)methanamine 95%
Dodavatel: Apollo Scientific
OR927518-1G
OR927518-250MG
APOSOR927518-1GEA
12010
CZK
APOSOR927518-1G
APOSOR927518-250MG
(4-Phenylmorpholin-2-yl)methanamine 95%
(4-Phenylmorpholin-2-yl)methanamine
Vzorec:
C₁₁H₁₆N₂O MW: 192,258 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06738826 CAS číslo: 112913-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...