Tisk…

7-Acetoxyindole 95%

Dodavatel: Apollo Scientific

OR927499-1G OR927499-250MG
APOSOR927499-1GEA 7010 CZK
APOSOR927499-1G APOSOR927499-250MG
7-Acetoxyindole 95%
7-Acetoxyindole
Vzorec: C₁₀H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00151953
CAS číslo: 5526-13-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR