Tisk…

Methyl 3-chloro-4-formylbenzoate 98%

Dodavatel: Apollo Scientific

OR927487-1G OR927487-5G
APOSOR927487-1GEA 2030 CZK
APOSOR927487-1G APOSOR927487-5G
Methyl 3-chloro-4-formylbenzoate 98%
Methyl 3-chloro-4-formylbenzoate
Vzorec: C₉H₇ClO₃
Storage Temperature: Ambient
MDL Number: MFCD16036862
CAS číslo: 74733-26-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR