Tisk…

5-Formyl-3-pyridinecarbonitrile 96%

Dodavatel: Apollo Scientific

Synonyms: 5-Formylnicotinonitrile

OR927407-1G OR927407-250MG
APOSOR927407-1GEA 21920 CZK
APOSOR927407-1G APOSOR927407-250MG
5-Formyl-3-pyridinecarbonitrile 96%
5-Formyl-3-pyridinecarbonitrile
Vzorec: C₇H₄N₂O
MW: 132,119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367908
CAS číslo: 70416-53-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR