2,5-Dichloroquinoxaline 95%
Dodavatel: Apollo Scientific
OR927312-1G
OR927312-250MG
APOSOR927312-1GEA
1870
CZK
APOSOR927312-1G
APOSOR927312-250MG
2,5-Dichloroquinoxaline 95%
2,5-Dichloroquinoxaline
Vzorec:
C₈H₄Cl₂N₂ MW: 199,037 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11846477 CAS číslo: 55687-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...