Tisk…

2-Pyridin-2-ylpyrimidine-5-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR927307-1G OR927307-250MG OR927307-5G
APOSOR927307-1GEA 6220 CZK
APOSOR927307-1G APOSOR927307-250MG APOSOR927307-5G
2-Pyridin-2-ylpyrimidine-5-carbaldehyde 95%
2-Pyridin-2-ylpyrimidine-5-carbaldehyde
Vzorec: C₁₀H₇N₃O
MW: 185.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09863209
CAS číslo: 954226-94-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR