5-(Pyridin-4-yl)-2H-1,2,4-triazol-3-ol 95%
Dodavatel: Apollo Scientific
OR927233-100MG
OR927233-1G
OR927233-250MG
APOSOR927233-100MGEA
4370
CZK
APOSOR927233-100MG
APOSOR927233-1G
APOSOR927233-250MG
5-(Pyridin-4-yl)-2H-1,2,4-triazol-3-ol 95%
5-(Pyridin-4-yl)-2H-1,2,4-triazol-3-ol
Vzorec:
C₇H₆N₄O Storage Temperature: Ambient |
MDL Number:
MFCD22565624 CAS číslo: 939-08-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...