Tisk…

4,5,6,7-Tetrahydro-1-benzothiophene-3-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR927126-1G OR927126-250MG
APOSOR927126-1GEA 16340 CZK
APOSOR927126-1G APOSOR927126-250MG
4,5,6,7-Tetrahydro-1-benzothiophene-3-carbaldehyde 95%
4,5,6,7-Tetrahydro-1-benzothiophene-3-carbaldehyde
Vzorec: C₉H₁₀OS
MW: 166,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12827456
CAS číslo: 851634-60-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR