Tisk…
(2,6,6-Trimethyl-2-hydroxycyclohexylidene)acetic acid lactone 95%
Dodavatel: Apollo Scientific
Synonyms: (R)-4,4,7a-Trimethyl-5,6,7,7a-tetrahydrobenzofuran-2(4H)-one, Dihydroactinidiolide
OR927125-1G OR927125-5G
APOSOR927125-1GEA 870 CZK
APOSOR927125-1G APOSOR927125-5G
(2,6,6-Trimethyl-2-hydroxycyclohexylidene)acetic acid lactone 95%
(2,6,6-Trimethyl-2-hydroxycyclohexylidene)acetic acid lactone
Vzorec: C₁₁H₁₆O₂
MW: 180,244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06409997
CAS číslo: 17092-92-1