5-Methyl-1H-pyrrole-3-carboxylic acid 97%
Dodavatel: Apollo Scientific
OR927108-1G
OR927108-250MG
APOSOR927108-1GEA
6190
CZK
APOSOR927108-1G
APOSOR927108-250MG
5-Methyl-1H-pyrrole-3-carboxylic acid 97%
5-Methyl-1H-pyrrole-3-carboxylic acid
Vzorec:
C₆H₇NO₂ MW: 125,125 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09991919 CAS číslo: 100047-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...