Tisk…

(2,6-Dichloro-phenyl)-ethyl-amine 95%

Dodavatel: Apollo Scientific

OR927073-1G OR927073-250MG
APOSOR927073-1GEA 9330 CZK
APOSOR927073-1G APOSOR927073-250MG
(2,6-Dichloro-phenyl)-ethyl-amine 95%
(2,6-Dichloro-phenyl)-ethyl-amine
Vzorec: C₈H₉Cl₂N
MW: 190.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658424
CAS číslo: 885280-60-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR