Tisk…

2-Chloro-5-phenyloxazole 97%

Dodavatel: Apollo Scientific

OR927050-100MG OR927050-250MG
APOSOR927050-100MGEA 2610 CZK
APOSOR927050-100MG APOSOR927050-250MG
2-Chloro-5-phenyloxazole 97%
2-Chloro-5-phenyloxazole
Vzorec: C₉H₆ClNO
MW: 179,603 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08688603
CAS číslo: 62124-43-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR