5-Chloro-N-methylpyridin-2-amine 98%
Dodavatel: Apollo Scientific
OR927031-1G
OR927031-5G
APOSOR927031-1GEA
1210
CZK
APOSOR927031-1G
APOSOR927031-5G
5-Chloro-N-methylpyridin-2-amine 98%
5-Chloro-N-methylpyridin-2-amine
Vzorec:
C₆H₇ClN₂ MW: 142.586 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00955783 CAS číslo: 4214-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...