Tisk…

3-Acetyl-5-chlorothiophene 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(5-Chlorothiophen-3-yl)ethanone

OR927023-1G OR927023-250MG
APOSOR927023-1GEA 1740 CZK
APOSOR927023-1G APOSOR927023-250MG
3-Acetyl-5-chlorothiophene 95%
3-Acetyl-5-chlorothiophene
Vzorec: C₆H₅ClOS
MW: 160,621 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082792
CAS číslo: 58119-67-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR