Tisk…

4-Chloro-2-(methylamino)benzoic acid 95%

Dodavatel: Apollo Scientific

OR926983-1G OR926983-250MG
APOSOR926983-1GEA 12400 CZK
APOSOR926983-1G APOSOR926983-250MG
4-Chloro-2-(methylamino)benzoic acid 95%
4-Chloro-2-(methylamino)benzoic acid
Vzorec: C₈H₈ClNO₂
MW: 185,608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17290555
CAS číslo: 54675-18-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR