Tisk…

Dimethyl ethylidenemalonate 95%

Dodavatel: Apollo Scientific

OR926979-10G OR926979-1G OR926979-5G
APOSOR926979-10GEA 8200 CZK
APOSOR926979-10G APOSOR926979-1G APOSOR926979-5G
Dimethyl ethylidenemalonate 95%
Dimethyl ethylidenemalonate
Vzorec: C₇H₁₀O₄
MW: 158,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008447
CAS číslo: 17041-60-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR