Tisk…

4-(tert-Butoxy)-2-chloropyrimidine 95%

Dodavatel: Apollo Scientific

OR926978-1G OR926978-250MG
APOSOR926978-1GEA 9330 CZK
APOSOR926978-1G APOSOR926978-250MG
4-(tert-Butoxy)-2-chloropyrimidine 95%
4-(tert-Butoxy)-2-chloropyrimidine
Vzorec: C₈H₁₁ClN₂O
MW: 186.639 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16712391
CAS číslo: 614729-28-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR