2,4,8-Trichloroquinazoline 97%
Dodavatel: Apollo Scientific
OR926964-1G
OR926964-250MG
OR926964-5G
APOSOR926964-1GEA
2070
CZK
APOSOR926964-1G
APOSOR926964-250MG
APOSOR926964-5G
2,4,8-Trichloroquinazoline 97%
2,4,8-Trichloroquinazoline
Vzorec:
C₈H₃Cl₃N₂ MW: 233,482 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09954870 CAS číslo: 62484-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...