Tisk…

(2,3-Dihydro-indol-1-yl)-acetic acid methyl ester 95%

Dodavatel: Apollo Scientific

OR926782-1G OR926782-250MG
APOSOR926782-1GEA 14290 CZK
APOSOR926782-1G APOSOR926782-250MG
(2,3-Dihydro-indol-1-yl)-acetic acid methyl ester 95%
(2,3-Dihydro-indol-1-yl)-acetic acid methyl ester
Vzorec: C₁₁H₁₃NO₂
MW: 191.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05147568
CAS číslo: 39597-68-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR