Tisk…

(1H-Imidazol-2-yl)methylamine

Dodavatel: Apollo Scientific

OR926767-1G OR926767-5G
APOSOR926767-1GEA 8870 CZK
APOSOR926767-1G APOSOR926767-5G
(1H-Imidazol-2-yl)methylamine
(1H-Imidazol-2-yl)methylamine
Vzorec: C₄H₇N₃
MW: 97,118 g/mol
Bod varu: 359 °C
Bod tání: 267…271 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114001
CAS číslo: 53332-80-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR