Tisk…
(R)-Piperidin-3-ylmethanol hydrochloride 97%
Dodavatel: Apollo Scientific
Synonyms: (R)-3-Piperidinemethanol hydrochloride
OR926711-1G OR926711-5G
APOSOR926711-1GEA 2070 CZK
APOSOR926711-1G APOSOR926711-5G
(R)-Piperidin-3-ylmethanol hydrochloride 97%
(R)-Piperidin-3-ylmethanol hydrochloride
Vzorec: C₆H₁₃NO·ClH
MW: 151,634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14155680
CAS číslo: 1124199-58-1