Tisk…

tert-Butyl N-ethyl-N-(2-hydroxyethyl)carbamate 98%

Dodavatel: Apollo Scientific

OR926561-25G OR926561-5G
APOSOR926561-25GEA 10020 CZK
APOSOR926561-25G APOSOR926561-5G
tert-Butyl N-ethyl-N-(2-hydroxyethyl)carbamate 98%
tert-Butyl N-ethyl-N-(2-hydroxyethyl)carbamate
Vzorec: C₉H₁₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD04973117
CAS číslo: 152192-95-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR