Tisk…

1-(2-Furylmethyl)-1H-pyrrole-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR926510-1G OR926510-5G
APOSOR926510-1GEA 4770 CZK
APOSOR926510-1G APOSOR926510-5G
1-(2-Furylmethyl)-1H-pyrrole-2-carbaldehyde 95%
1-(2-Furylmethyl)-1H-pyrrole-2-carbaldehyde
Vzorec: C₁₀H₉NO₂
MW: 175,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09674985
CAS číslo: 13788-32-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR