Tisk…
1-Chloro-2-methylpropyl propionate 95%
Dodavatel: Apollo Scientific
OR926388-1G OR926388-5G
APOSOR926388-1GEA 1080 CZK
APOSOR926388-1G APOSOR926388-5G
1-Chloro-2-methylpropyl propionate 95%
1-Chloro-2-methylpropyl propionate
Vzorec: C₇H₁₃ClO₂
MW: 164.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09033184
CAS číslo: 58304-65-7