Tisk…

2-(4-Hydroxyphenyl)-2-methylpropanoic acid 95%

Dodavatel: Apollo Scientific

OR926346-1G OR926346-250MG
APOSOR926346-1GEA 4930 CZK
APOSOR926346-1G APOSOR926346-250MG
2-(4-Hydroxyphenyl)-2-methylpropanoic acid 95%
2-(4-Hydroxyphenyl)-2-methylpropanoic acid
Vzorec: C₁₀H₁₂O₃
MW: 180,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00995712
CAS číslo: 29913-51-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR