Tisk…

2-Chloro-5-methylthiazole-4-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR926189-1G OR926189-250MG
APOSOR926189-1GEA 1950 CZK
APOSOR926189-1G APOSOR926189-250MG
2-Chloro-5-methylthiazole-4-carboxylic acid 95%
2-Chloro-5-methylthiazole-4-carboxylic acid
Vzorec: C₅H₄ClNO₂S
Storage Temperature: Ambient
MDL Number: MFCD12404922
CAS číslo: 1194374-24-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR