Tisk…

Pyridin-2-yl-acetic acid tert-butyl ester 95%

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl 2-(pyridin-2-yl)acetate

OR926158-1G OR926158-250MG
APOSOR926158-1GEA 10190 CZK
APOSOR926158-1G APOSOR926158-250MG
Pyridin-2-yl-acetic acid tert-butyl ester 95%
Pyridin-2-yl-acetic acid tert-butyl ester
Vzorec: C₁₁H₁₅NO₂
MW: 193,242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19441884
CAS číslo: 150059-62-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR