Tisk…

(8-Methylimidazo[1,2-a]pyridin-2-yl)methanol 95%

Dodavatel: Apollo Scientific

OR926138-1G OR926138-250MG
APOSOR926138-1GEA 10350 CZK
APOSOR926138-1G APOSOR926138-250MG
(8-Methylimidazo[1,2-a]pyridin-2-yl)methanol 95%
(8-Methylimidazo[1,2-a]pyridin-2-yl)methanol
Vzorec: C₉H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD08752621
CAS číslo: 872362-98-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR