3-Pyridin-3-yl-[1,2,4]oxadiazol-5-ol 95%
Dodavatel: Apollo Scientific
OR926110-1G
OR926110-250MG
APOSOR926110-1GEA
14290
CZK
APOSOR926110-1G
APOSOR926110-250MG
3-Pyridin-3-yl-[1,2,4]oxadiazol-5-ol 95%
3-Pyridin-3-yl-[1,2,4]oxadiazol-5-ol
Vzorec:
C₇H₅N₃O₂ MW: 163,134 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19441961 CAS číslo: 1198-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...