Tisk…

7-Methyl-DL-tryptophan 97%

Dodavatel: Apollo Scientific

Synonyms: H-DL-Trp(7-Me)-OH, 2-Amino-3-(7-methyl-1H-indol-3-yl)propanoic acid

OR926023-1G OR926023-250MG
APOSOR926023-1GEA 5460 CZK
APOSOR926023-1G APOSOR926023-250MG
7-Methyl-DL-tryptophan 97%
7-Methyl-DL-tryptophan
Vzorec: C₁₂H₁₄N₂O₂
MW: 218.252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00056926
CAS číslo: 17332-70-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR