5-(1H-Pyrazol-5-yl)-2-furaldehyde 95%
Dodavatel: Apollo Scientific
OR925945-1G
OR925945-5G
APOSOR925945-1GEA
6220
CZK
APOSOR925945-1G
APOSOR925945-5G
5-(1H-Pyrazol-5-yl)-2-furaldehyde 95%
5-(1H-Pyrazol-5-yl)-2-furaldehyde
Vzorec:
C₈H₆N₂O₂ MW: 162,145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08059993 CAS číslo: 1015939-90-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...