Tisk…
5-Methoxyisoquinoline hydrochloride 95%
Dodavatel: Apollo Scientific
OR925755-5G
APOSOR925755-5GEA 4600 CZK
APOSOR925755-5G
5-Methoxyisoquinoline hydrochloride 95%
5-Methoxyisoquinoline hydrochloride
Vzorec: C₁₀H₉NO·ClH
MW: 195,646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22683320
CAS číslo: 1418117-87-9