2-Chloro-4-hydroxypyrimidine 95%
Dodavatel: Apollo Scientific
Synonyms:
2-Chloropyrimidin-4-ol
OR925753-1G
OR925753-250MG
APOSOR925753-1GEA
1870
CZK
APOSOR925753-1G
APOSOR925753-250MG
2-Chloro-4-hydroxypyrimidine 95%
2-Chloro-4-hydroxypyrimidine
Vzorec:
C₄H₃ClN₂O MW: 130,532 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09038284 CAS číslo: 55873-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...