Tisk…

1-(2-Methoxymethoxy-phenyl)-ethanone 95%

Dodavatel: Apollo Scientific

OR925660-1G OR925660-5G
APOSOR925660-1GEA 6620 CZK
APOSOR925660-1G APOSOR925660-5G
1-(2-Methoxymethoxy-phenyl)-ethanone 95%
1-(2-Methoxymethoxy-phenyl)-ethanone
Vzorec: C₁₀H₁₂O₃
MW: 180,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20921560
CAS číslo: 6515-18-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR